1,5-Dichloro-2,4-difluorobenzene 97%

Supplier: Ambeed
Warning

Synonyms: 1,3-Dichloro-4,6-difluorobenzene

A494318-1G A494318-250MG
76991-174EA 89.12 USD
76991-174 76991-176
1,5-Dichloro-2,4-difluorobenzene 97%
1,5-Dichloro-2,4-difluorobenzene

Precaución: for Research Use Only

Formula: C₆H₂Cl₂F₂
MW: 182.98 g/mol
MDL Number: MFCD11226309
CAS Number: 2253-30-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR