N-((2S,3R,4S,5R)-3,4,5,6-Tetrahydroxy-1-oxohexan-2-yl)acetamide 98%

Supplier: Ambeed
Warning

A494199-1G A494199-250MG A494199-5G
76991-168EA 61.54 USD
76991-168 76991-170 76991-172
N-((2S,3R,4S,5R)-3,4,5,6-Tetrahydroxy-1-oxohexan-2-yl)acetamide 98%
N-((2S,3R,4S,5R)-3,4,5,6-Tetrahydroxy-1-oxohexan-2-yl)acetamide

Precaución: for Research Use Only

Formula: C₈H₁₅NO₆
MW: 221.208 g/mol
MDL Number: MFCD00046758
CAS Number: 3615-17-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR