5-Chloroisoquinolin-1(2H)-one 95%

Supplier: Ambeed
Warning

A494086-100MG A494086-1G A494086-250MG
76991-162EA 9 USD
76991-162 76991-164 76991-166
5-Chloroisoquinolin-1(2H)-one 95%
5-Chloroisoquinolin-1(2H)-one

Precaución: for Research Use Only

Formula: C₉H₆ClNO
MDL Number: MFCD11847594
CAS Number: 24188-73-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR