(1-Ethyl-1H-pyrazol-4-yl)methanol 97%

Supplier: Ambeed
Warning

A493747-1G A493747-250MG
77034-428EA 97.61 USD
77034-428 77034-430
(1-Ethyl-1H-pyrazol-4-yl)methanol 97%
(1-Ethyl-1H-pyrazol-4-yl)methanol

Precaución: for Research Use Only

Formula: C₆H₁₀N₂O
MW: 126.16 g/mol
MDL Number: MFCD02253733
CAS Number: 905307-04-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR