4-(Hydrazinocarbonyl)benzeneboronic acid pinacol ester 98%

Supplier: Ambeed
Danger

Synonyms: 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzohydrazide

A493738-1G A493738-250MG A493738-5G
77017-170EA 57 USD
77017-170 77017-172 77017-174
4-(Hydrazinocarbonyl)benzeneboronic acid pinacol ester 98%
4-(Hydrazinocarbonyl)benzeneboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₉BN₂O₃
MDL Number: MFCD07783860
CAS Number: 276694-16-7
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR