Methyl 3-bromo-4-cyclohexylbenzoate 95+%
Supplier: Ambeed
|
Warning
|
A493235-1G
A493235-250MG
76991-150EA
69
USD
76991-150
76991-152
Methyl 3-bromo-4-cyclohexylbenzoate 95+%
Methyl 3-bromo-4-cyclohexylbenzoate
Precaución: for Research Use Only
Formula:
C₁₄H₁₇BrO₂ MW: 297.18 g/mol |
MDL Number:
MFCD11113008 CAS Number: 1131594-38-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...