1-Boc-3-Hydroxy-4-methylpiperidine 98%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 3-hydroxy-4-methylpiperidine-1-carboxylate

A493156-100MG A493156-1G A493156-250MG
77034-422EA 132.62 USD
77034-422 77034-424 77034-426
1-Boc-3-Hydroxy-4-methylpiperidine 98%
1-Boc-3-Hydroxy-4-methylpiperidine

Precaución: for Research Use Only

Formula: C₁₁H₂₁NO₃
MW: 215.28 g/mol
MDL Number: MFCD13152263
CAS Number: 1188264-78-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR