2-Bromo-4-methyl-1-nitrobenzene 98%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-4-nitrotoluene

A492711-10G A492711-1G A492711-250MG A492711-25G A492711-5G
76991-140EA 140 USD
76991-140 76991-142 76991-144 76991-146 76991-148
2-Bromo-4-methyl-1-nitrobenzene 98%
2-Bromo-4-methyl-1-nitrobenzene

Precaución: for Research Use Only

Formula: C₇H₆BrNO₂
MDL Number: MFCD07363742
CAS Number: 40385-54-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR