2-(N-(2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)ethyl)-2-(4-(((benzhydryloxy)carbonyl)amino)-2-oxopyrimidin-1(2H)-yl)acetamido)acetic acid ≥95%, Ambeed.Inc
Supplier: Ambeed
|
Warning
|
Synonyms:
Fmoc-PNA-C(Bhoc)-OH
A492482-100MG
A492482-250MG
A492482-1G
A492482-5G
76991-130EA
55
USD
76991-130
76991-132
77613-554
77684-314
2-(N-(2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)ethyl)-2-(4-(((benzhydryloxy)carbonyl)amino)-2-oxopyrimidin-1(2H)-yl)acetamido)acetic acid ≥95%, Ambeed.Inc
2-(N-(2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)ethyl)-2-(4-(((benzhydryloxy)carbonyl)amino)-2-oxopyrimidin-1(2H)-yl)acetamido)acetic acid
Precaución: For research use only.
Formula:
C₃₉H₃₅N₅O₈ MW: 701.72 g/mol |
MDL Number:
MFCD22373799 CAS Number: 186046-81-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...