4-Isobutylphenol 97%

Supplier: Ambeed
Warning

A492402-1G A492402-250MG
77017-166EA 481.65 USD
77017-166 77017-168
4-Isobutylphenol 97%
4-Isobutylphenol

Precaución: for Research Use Only

Formula: C₁₀H₁₄O
MW: 150.218 g/mol
MDL Number: MFCD01846454
CAS Number: 4167-74-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR