3-Bromo-5-chloro-6-methylpyridin-2-amine 98%

Supplier: Ambeed
Warning

A491810-100MG A491810-1G A491810-250MG A491810-5G
77017-150EA 12 USD
77017-150 77017-152 77017-154 77017-156
3-Bromo-5-chloro-6-methylpyridin-2-amine 98%
3-Bromo-5-chloro-6-methylpyridin-2-amine

Precaución: for Research Use Only

Formula: C₆H₆BrClN₂
MW: 221.48 g/mol
MDL Number: MFCD09754393
CAS Number: 952948-94-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR