[2,2'-Bipyridin]-5-amine 95%

Supplier: Ambeed
Warning

A491273-100MG A491273-250MG A491273-1G
76991-122EA 46.68 USD
76991-122 76991-124 77613-548
[2,2'-Bipyridin]-5-amine 95%
[2,2'-Bipyridin]-5-amine

Precaución: for Research Use Only

Formula: C₁₀H₉N₃
MW: 171.19 g/mol
MDL Number: MFCD12033465
CAS Number: 160539-04-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR