8-Methylquinazoline 95%

Supplier: Ambeed
Warning

A491262-1G A491262-250MG
77017-146EA 301 USD
77017-146 77017-148
8-Methylquinazoline 95%
8-Methylquinazoline

Precaución: for Research Use Only

Formula: C₉H₈N₂
MDL Number: MFCD18448927
CAS Number: 7557-03-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR