2-Fluoro-N-methyl-4-nitrobenzamide 95%

Supplier: Ambeed
Warning

A490981-10G A490981-1G A490981-5G
76991-108EA 148.53 USD
76991-108 76991-110 76991-112
2-Fluoro-N-methyl-4-nitrobenzamide 95%
2-Fluoro-N-methyl-4-nitrobenzamide

Precaución: for Research Use Only

Formula: C₈H₇FN₂O₃
MW: 198.151 g/mol
MDL Number: MFCD09909407
CAS Number: 915087-24-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR