4'-(4-Methoxyphenyl)-2,2':6',2''-terpyridine 97%

Supplier: Ambeed
Warning

A490839-100MG A490839-1G A490839-5G A490839-250MG A490839-25G
77017-132EA 14 USD
77017-132 77017-134 77017-136 77613-544 77890-997
4'-(4-Methoxyphenyl)-2,2':6',2''-terpyridine 97%
4'-(4-Methoxyphenyl)-2,2':6',2''-terpyridine

Precaución: for Research Use Only

Formula: C₂₂H₁₇N₃O
MDL Number: MFCD06796987
CAS Number: 13104-56-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR