4-(Trifluoromethyl)-3-pyridineboronic acid pinacol ester 96%

Supplier: Ambeed
Warning

Synonyms: 4-(Trifluoromethyl)pyridine-3-boronic acid pinacol ester, 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-4-(trifluoromethyl)pyridine

A490484-1G A490484-250MG A490484-5G
76991-102EA 66 USD
76991-102 76991-104 76991-106
4-(Trifluoromethyl)-3-pyridineboronic acid pinacol ester 96%
4-(Trifluoromethyl)-3-pyridineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₅BF₃NO₂
MDL Number: MFCD12198140
CAS Number: 1310405-06-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR