3-Bromo-1-methylpyridin-4(1H)-one 95%
Supplier: Ambeed
|
Warning
|
A490367-250MG
A490367-5G
77017-124EA
39.26
USD
77017-124
77684-305
3-Bromo-1-methylpyridin-4(1H)-one 95%
3-Bromo-1-methylpyridin-4(1H)-one
Precaución: For research use only.
Formula:
C₆H₆BrNO MW: 188.02 g/mol |
MDL Number:
MFCD23161474 CAS Number: 87875-76-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...