(5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)thiophen-2-yl)methanol 98%, Ambeed.Inc

Supplier: Ambeed
Warning

A490249-100MG A490249-250MG A490249-1G A490249-5G
77017-120EA 12 USD
77017-120 77017-122 77684-304 77890-995
(5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)thiophen-2-yl)methanol 98%, Ambeed.Inc
(5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)thiophen-2-yl)methanol

Precaución: For research use only.

Formula: C₁₁H₁₇BO₃S
MDL Number: MFCD18383085
CAS Number: 1026796-39-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR