Diethyl-2-propynylmalonate 96%

Supplier: Ambeed
Warning

Synonyms: 2-Propynylmalonic Acid Diethyl Ester, Diethyl Propargylmalonate, Propargylmalonic Acid Diethyl Ester, Diethyl 2-(prop-2-yn-1-yl)malonate

A489985-1G A489985-250MG A489985-5G
76991-096EA 50.93 USD
76991-096 76991-098 76991-100
Diethyl-2-propynylmalonate 96%
Diethyl-2-propynylmalonate

Precaución: for Research Use Only

Formula: C₁₀H₁₄O₄
MW: 198.22 g/mol
MDL Number: MFCD00091967
CAS Number: 17920-23-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR