5-Fluoro-2-indoleboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 5-Fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole

A489521-100MG A489521-1G A489521-250MG A489521-5G
77017-112EA 34 USD
77017-112 77017-114 77017-116 77017-118
5-Fluoro-2-indoleboronic acid pinacol ester 98%
5-Fluoro-2-indoleboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₁₇BFNO₂
MDL Number: MFCD11858362
CAS Number: 1256358-94-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR