(R,R)-(-)-N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediaminomanganese(III) chloride 98%

Supplier: Ambeed
Warning

A489410-1G A489410-25G A489410-5G A489410-100G
77034-408EA 7 USD
77034-408 77034-410 77034-412 77872-810
(R,R)-(-)-N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediaminomanganese(III) chloride 98%
(R,R)-(-)-N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediaminomanganese(III) chloride

Precaución: for Research Use Only

Formula: C₃₆H₅₂ClMnN₂O₂
MDL Number: MFCD02101664
CAS Number: 138124-32-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR