2-Oxo-2,3-dihydrobenzo[d]oxazole-5-sulfonyl chloride 98%

Supplier: Ambeed
Danger

A489357-100MG A489357-250MG A489357-1G
77017-108EA 32 USD
77017-108 77017-110 77568-958
2-Oxo-2,3-dihydrobenzo[d]oxazole-5-sulfonyl chloride 98%
2-Oxo-2,3-dihydrobenzo[d]oxazole-5-sulfonyl chloride

Precaución: For research use only.

Formula: C₇H₄ClNO₄S
MDL Number: MFCD04629624
CAS Number: 78633-41-7
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR