4-Bromo-7-chloro-1H-pyrrolo[3,2-c]pyridine 95%

Supplier: Ambeed
Warning

A489298-100MG A489298-1G A489298-250MG
77017-102EA 234.46 USD
77017-102 77017-104 77017-106
4-Bromo-7-chloro-1H-pyrrolo[3,2-c]pyridine 95%
4-Bromo-7-chloro-1H-pyrrolo[3,2-c]pyridine

Precaución: for Research Use Only

Formula: C₇H₄BrClN₂
MW: 231.48 g/mol
MDL Number: MFCD09749991
CAS Number: 1000341-81-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR