N,N'-Bis(5-hydroxysalicylidene)ethylenediamine 98%

Supplier: Ambeed
Warning

Synonyms: N,N'-Ethylenebis(5-hydroxysalicylideneimine), N,N-Bis(2,5-dihydroxybenzylidene)ethylenediamine, 2,2'-((Ethane-1,2-diylbis(azanylylidene))bis(methanylylidene))bis(benzene-1,4-diol)

A489149-1G A489149-250MG A489149-5G A489149-25G
77034-402EA 38 USD
77034-402 77034-404 77034-406 77872-809
N,N'-Bis(5-hydroxysalicylidene)ethylenediamine 98%
N,N'-Bis(5-hydroxysalicylidene)ethylenediamine

Precaución: for Research Use Only

Formula: C₁₆H₁₆N₂O₄
MW: 300.31 g/mol
MDL Number: MFCD04039747
CAS Number: 124061-43-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR