5-Methyl-2-(methylthio)pyrimidin-4(3H)-one 97%

Supplier: Ambeed
Warning

A488420-10G A488420-1G A488420-250MG A488420-5G
76991-088EA 500.75 USD
76991-088 76991-090 76991-092 77684-299
5-Methyl-2-(methylthio)pyrimidin-4(3H)-one 97%
5-Methyl-2-(methylthio)pyrimidin-4(3H)-one

Precaución: For research use only.

Formula: C₆H₈N₂OS
MW: 156.21 g/mol
MDL Number: MFCD00194961
CAS Number: 20651-30-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR