H-Orn(2-Cl-Z)-OH 95%
Supplier: Ambeed
|
Warning
|
A488406-10G
A488406-1G
A488406-5G
77004-618EA
15.92
USD
77004-618
77004-620
77004-622
H-Orn(2-Cl-Z)-OH 95%
H-Orn(2-Cl-Z)-OH
Precaución: for Research Use Only
Formula:
C₁₃H₁₇ClN₂O₄ MW: 300.8 g/mol |
MDL Number:
MFCD00235874 CAS Number: 118553-99-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...