(S)-3-Cyclohexylmorpholine 95%

Supplier: Ambeed
Warning

A488360-100MG A488360-1G A488360-250MG
76991-082EA 77.45 USD
76991-082 76991-084 76991-086
(S)-3-Cyclohexylmorpholine 95%
(S)-3-Cyclohexylmorpholine

Precaución: for Research Use Only

Formula: C₁₀H₁₉NO
MW: 169.26 g/mol
MDL Number: MFCD18665124
CAS Number: 1270289-34-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR