4-(3-Aminophenyl)morpholin-3-one 95%
Supplier: Ambeed
|
Warning
|
A488232-100MG
A488232-250MG
76991-070EA
169.75
USD
76991-070
76991-072
4-(3-Aminophenyl)morpholin-3-one 95%
4-(3-Aminophenyl)morpholin-3-one
Precaución: for Research Use Only
Formula:
C₁₀H₁₂N₂O₂ MW: 192.21 g/mol |
MDL Number:
MFCD11577324 CAS Number: 1082495-22-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...