Methyl 6-(trifluoromethyl)pyridine-2-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 6-(trifluoromethyl)picolinate, Methyl 6-(trifluoromethyl)picolinate

A487868-10G A487868-1G A487868-5G
77034-396EA 53 USD
77034-396 77034-398 77034-400
Methyl 6-(trifluoromethyl)pyridine-2-carboxylate 97%
Methyl 6-(trifluoromethyl)pyridine-2-carboxylate

Precaución: for Research Use Only

Formula: C₈H₆F₃NO₂
MW: 205.134 g/mol
MDL Number: MFCD02090527
CAS Number: 155377-05-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR