1,3-Bis(2,6-diisopropylphenyl)imidazolium tetrafluoroborate 98%

Supplier: Ambeed
Warning

A487803-10G A487803-1G A487803-250MG A487803-5G
76991-060EA 218 USD
76991-060 76991-062 76991-064 76991-066
1,3-Bis(2,6-diisopropylphenyl)imidazolium tetrafluoroborate 98%
1,3-Bis(2,6-diisopropylphenyl)imidazolium tetrafluoroborate

Precaución: for Research Use Only

Formula: C₂₇H₃₇BF₄N₂
MDL Number: MFCD04971187
CAS Number: 286014-25-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR