5-Amino-2-iodopyridine 98%

Supplier: Ambeed
Warning

A486681-5G A486681-250MG
77004-606EA 132 USD
77004-606 77882-223
5-Amino-2-iodopyridine 98%
5-Amino-2-iodopyridine

Precaución: for Research Use Only

Formula: C₅H₅IN₂
MDL Number: MFCD02706663
CAS Number: 29958-12-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR