2-Chloro-6-(trifluoromethyl)pyridin-4-amine 96%

Supplier: Ambeed
Warning

A486614-250MG A486614-25G
76991-056EA 37.14 USD
76991-056 76991-058
2-Chloro-6-(trifluoromethyl)pyridin-4-amine 96%
2-Chloro-6-(trifluoromethyl)pyridin-4-amine

Precaución: for Research Use Only

Formula: C₆H₄ClF₃N₂
MW: 196.55 g/mol
MDL Number: MFCD12828259
CAS Number: 34486-22-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR