(1S,2S)-N,N'-Bis[2-(diphenylphosphino)benzyl]cyclohexane-1,2-diamine ≥98%, ee 99%

Supplier: Ambeed
Warning

Synonyms: (1S,2S)-N1,N1-Bis(2-(diphenylphosphino)benzyl)cyclohexane-1,2-diamine

A486402-100MG A486402-1G A486402-250MG A486402-5G
76991-048EA 37 USD
76991-048 76991-050 76991-052 76991-054
(1S,2S)-N,N'-Bis[2-(diphenylphosphino)benzyl]cyclohexane-1,2-diamine ≥98%, ee 99%
(1S,2S)-N,N'-Bis[2-(diphenylphosphino)benzyl]cyclohexane-1,2-diamine

Precaución: for Research Use Only

Formula: C₄₄H₄₄N₂P₂
MW: 662.798 g/mol
MDL Number: MFCD17014022
CAS Number: 174677-83-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR