2,4-Dichloro-7-(trifluoromethyl)quinazoline 98%

Supplier: Ambeed
Warning

A485625-1G A485625-250MG A485625-5G
77034-382EA 116 USD
77034-382 77034-384 77034-386
2,4-Dichloro-7-(trifluoromethyl)quinazoline 98%
2,4-Dichloro-7-(trifluoromethyl)quinazoline

Precaución: for Research Use Only

Formula: C₉H₃Cl₂F₃N₂
MW: 267.0347 g/mol
MDL Number: MFCD09954878
CAS Number: 396-02-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR