4-Bromo-1-benzenesulfonyl-7-azaindole 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-Benzenesulphonyl-4-bromo-7-azaindole
A485379-1G
A485379-250MG
A485379-5G
76991-038EA
93.36
USD
76991-038
76991-040
76991-042
4-Bromo-1-benzenesulfonyl-7-azaindole 98%
4-Bromo-1-benzenesulfonyl-7-azaindole
Precaución: for Research Use Only
Formula:
C₁₃H₉BrN₂O₂S MW: 337.2 g/mol |
MDL Number:
MFCD08443800 CAS Number: 889939-25-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...