Methyl-4-aminopyrimidine-5-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-aminopyrimidine-5-carboxylate

A485327-1G A485327-250MG
77034-378EA 119.89 USD
77034-378 77034-380
Methyl-4-aminopyrimidine-5-carboxylate 98%
Methyl-4-aminopyrimidine-5-carboxylate

Precaución: for Research Use Only

Formula: C₆H₇N₃O₂
MW: 153.1400 g/mol
MDL Number: MFCD13195321
CAS Number: 714975-53-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR