(R)-(-)-8-Diphenylphosphino-1,2,3,4-tetrahydro-1-naphthylamine 98%

Supplier: Ambeed
Warning

A484911-100MG A484911-1G A484911-250MG A484911-5G
76991-030EA 36 USD
76991-030 76991-032 76991-034 76991-036
(R)-(-)-8-Diphenylphosphino-1,2,3,4-tetrahydro-1-naphthylamine 98%
(R)-(-)-8-Diphenylphosphino-1,2,3,4-tetrahydro-1-naphthylamine

Precaución: for Research Use Only

Formula: C₂₂H₂₂NP
MDL Number: MFCD16621442
CAS Number: 960128-64-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR