Methyl isoquinoline-5-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 5-Isoquinolinecarboxylate

A484655-100MG A484655-1G A484655-250MG A484655-5G
76991-022EA 29.71 USD
76991-022 76991-024 76991-026 76991-028
Methyl isoquinoline-5-carboxylate 98%
Methyl isoquinoline-5-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₉NO₂
MW: 187.195 g/mol
MDL Number: MFCD06204278
CAS Number: 16675-59-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR