Ethyl 4-((tert-butoxycarbonyl)amino)-1-methyl-1H-imidazole-2-carboxylate ≥98%

Supplier: Ambeed
Warning

A484508-100MG A484508-1G A484508-250MG
77017-092EA 5 USD
77017-092 77017-094 77017-096
Ethyl 4-((tert-butoxycarbonyl)amino)-1-methyl-1H-imidazole-2-carboxylate ≥98%
Ethyl 4-((tert-butoxycarbonyl)amino)-1-methyl-1H-imidazole-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₉N₃O₄
MDL Number: MFCD16876078
CAS Number: 128293-63-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR