8-(Trifluoromethoxy)quinoline-5-boronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (8-(Trifluoromethoxy)quinolin-5-yl)boronic acid

A484479-1G A484479-250MG A484479-5G
77017-086EA 210 USD
77017-086 77017-088 77017-090
8-(Trifluoromethoxy)quinoline-5-boronic acid 98%
8-(Trifluoromethoxy)quinoline-5-boronic acid

Precaución: for Research Use Only

Formula: C₁₀H₇BF₃NO₃
MDL Number: MFCD11504851
CAS Number: 1072951-46-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR