(E)-2-(3,5-Difluorostyryl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 96%
Supplier: Ambeed
|
Warning
|
A483960-100MG
A483960-1G
A483960-250MG
A483960-5G
76991-014EA
16
USD
76991-014
76991-016
76991-018
76991-020
(E)-2-(3,5-Difluorostyryl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 96%
(E)-2-(3,5-Difluorostyryl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Precaución: for Research Use Only
Formula:
C₁₄H₁₇BF₂O₂ |
MDL Number:
MFCD08457659 CAS Number: 1073354-58-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...