1-Cbz-4-(aminomethyl)piperidine 98%

Supplier: Ambeed
Warning

Synonyms: 1-Cbz-4-Aminomethylpiperidine

A483794-1G A483794-5G A483794-10G A483794-25G
77004-602EA 29 USD
77004-602 77004-604 77673-260 77673-262
1-Cbz-4-(aminomethyl)piperidine 98%
1-Cbz-4-(aminomethyl)piperidine

Precaución: for Research Use Only

Formula: C₁₄H₂₀N₂O₂
MW: 248.32 g/mol
MDL Number: MFCD03001690
CAS Number: 157023-34-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR