Ethyl (2-fluorobenzoyl)acetate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 3-(2-fluorophenyl)-3-oxo-propionate

A483531-1G A483531-250MG A483531-25G A483531-5G
77017-064EA 23.34 USD
77017-064 77017-066 77017-068 77017-070
Ethyl (2-fluorobenzoyl)acetate 98%
Ethyl (2-fluorobenzoyl)acetate

Precaución: for Research Use Only

Formula: C₁₁H₁₁FO₃
MW: 210.204 g/mol
MDL Number: MFCD00000321
CAS Number: 1479-24-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR