4-Fluoro-2-(methoxycarbonyl)benzeneboronic acid pinacol ester 95+%

Supplier: Ambeed
Warning

Synonyms: Methyl 5-fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

A483065-100MG A483065-1G A483065-250MG
77017-058EA 39 USD
77017-058 77017-060 77017-062
4-Fluoro-2-(methoxycarbonyl)benzeneboronic acid pinacol ester 95+%
4-Fluoro-2-(methoxycarbonyl)benzeneboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₁₈BFO₄
MW: 280.102 g/mol
MDL Number: MFCD16996305
CAS Number: 1293284-58-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR