7-Bromoheptan-1-amine hydrobromide 95%

Supplier: Ambeed
Danger

Synonyms: 7-Bromo-1-heptanamine Hydrobromide

A482862-250MG A482862-100MG A482862-1G
77017-052EA 333.13 USD
77017-052 77613-510 77684-281
7-Bromoheptan-1-amine hydrobromide 95%
7-Bromoheptan-1-amine hydrobromide

Precaución: For research use only.

Formula: C₇H₁₇Br₂N
MW: 275.02 g/mol
MDL Number: MFCD28118492
CAS Number: 1082678-45-2
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR