5-Bromo-2-(trifluoromethoxy)aniline 98%

Supplier: Ambeed
Warning

Synonyms: 3-Amino-4-(trifluoromethoxy)bromobenzene

A482365-100G A482365-10G A482365-25G A482365-500G A482365-5G
77034-352EA 69 USD
77034-352 77034-354 77034-356 77034-358 77034-360
5-Bromo-2-(trifluoromethoxy)aniline 98%
5-Bromo-2-(trifluoromethoxy)aniline

Precaución: for Research Use Only

Formula: C₇H₅BrF₃NO
MDL Number: MFCD04039242
CAS Number: 886762-08-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR