(2-Chlorophenyl)(pyridin-2-yl)methanone 95%

Supplier: Ambeed
Warning

A482284-100MG A482284-1G A482284-250MG
77017-040EA 40.32 USD
77017-040 77017-042 77017-044
(2-Chlorophenyl)(pyridin-2-yl)methanone 95%
(2-Chlorophenyl)(pyridin-2-yl)methanone

Precaución: for Research Use Only

Formula: C₁₂H₈ClNO
MW: 217.65 g/mol
MDL Number: MFCD07699234
CAS Number: 1694-57-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR