(R)-Quinuclidin-3-amine 98%
Supplier: Ambeed
|
Danger
|
A481670-250MG
A481670-100MG
76991-010EA
172.93
USD
76991-010
77684-279
(R)-Quinuclidin-3-amine 98%
(R)-Quinuclidin-3-amine
Precaución: For research use only.
Formula:
C₇H₁₄N₂ MW: 126.19 g/mol |
MDL Number:
MFCD08669630 CAS Number: 123536-15-2 UN: 3259 ADR: 8,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...