5,7-Difluoro-1H-indole-2-carboxylic acid 97%

Supplier: Ambeed
Warning

A481599-100G A481599-1G A481599-25G A481599-5G
77004-594EA 1242.32 USD
77004-594 77004-596 77004-598 77004-600
5,7-Difluoro-1H-indole-2-carboxylic acid 97%
5,7-Difluoro-1H-indole-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₅F₂NO₂
MW: 197.14 g/mol
MDL Number: MFCD01863161
CAS Number: 186432-20-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR