2-Hydroxy-4-methyl-6-(trifluoromethyl)nicotinamide 95+%

Supplier: Ambeed
Warning

A481308-100MG A481308-1G A481308-250MG
77004-588EA 18 USD
77004-588 77004-590 77004-592
2-Hydroxy-4-methyl-6-(trifluoromethyl)nicotinamide 95+%
2-Hydroxy-4-methyl-6-(trifluoromethyl)nicotinamide

Precaución: for Research Use Only

Formula: C₈H₇F₃N₂O₂
MDL Number: MFCD16659045
CAS Number: 116548-07-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR